Assay Design
Gene Transcripts
- Gene Symbol
- cand-1
- Entrez Gene ID
-
180125
- Gene Name
- Cullin-associated NEDD8-dissociated protein 1
- Gene Aliases
-
CELE_Y102A5A.1
- Chromosome Location
-
Chr.V: 16802872 - 16814957 on Build WBcel235
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_074843.6 |
NP_507244.1 |
1 - 2 |
145 |
68 |
View Full Product Details