Assay Design
Gene Transcripts
- Gene Symbol
- phy-3
- Entrez Gene ID
-
188624
- Gene Name
- Fe2OG dioxygenase domain-containing protein
- Gene Aliases
-
CELE_T20B3.7
- Chromosome Location
-
Chr.V: 16833165 - 16839141 on Build WBcel235
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_074850.3 |
NP_507251.2 |
3 - 4 |
583 |
98 |
View Full Product Details