Assay Design
Gene Transcripts
- Gene Symbol
- clec-26
- Entrez Gene ID
-
188626
- Gene Name
- C-type LECtin
- Gene Aliases
-
CELE_T20B3.12
- Chromosome Location
-
Chr.V: 16848611 - 16850199 on Build WBcel235
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_074853.4 |
NP_507254.2 |
5 - 6 |
831 |
124 |
View Full Product Details