Assay Design



Gene Transcripts

Gene Symbol
srz-66
Entrez Gene ID
185527
Gene Name
Serpentine Receptor, class Z
Gene Aliases
CELE_F40D4.9
Chromosome Location
Chr.V: 17166028 - 17167557 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001356779.1 NP_001343582.1 1 - 2 223 141
NR_002399.1 1 - 2 223 141

View Full Product Details