Assay Design
Gene Transcripts
- Gene Symbol
- srbc-25
- Entrez Gene ID
-
185524
- Gene Name
- Serpentine Receptor, class BC (class B-like)
- Gene Aliases
-
CELE_F40D4.6
- Chromosome Location
-
Chr.V: 17174038 - 17175530 on Build WBcel235
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_074945.1 |
NP_507346.1 |
5 - 6 |
750 |
137 |
View Full Product Details