Assay Design
Gene Transcripts
- Gene Symbol
- str-89
- Entrez Gene ID
-
186558
- Gene Name
- Seven TM Receptor
- Gene Aliases
-
CELE_F59A1.4
- Chromosome Location
-
Chr.V: 17668831 - 17671330 on Build WBcel235
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_075071.2 |
NP_507472.1 |
2 - 3 |
320 |
84 |
View Full Product Details