Assay Design
Gene Transcripts
- Gene Symbol
- str-77
- Entrez Gene ID
-
189247
- Gene Name
- Seven TM Receptor
- Gene Aliases
-
CELE_W06D12.4
- Chromosome Location
-
Chr.V: 17718947 - 17721293 on Build WBcel235
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_075080.1 |
NP_507481.1 |
4 - 5 |
783 |
78 |
View Full Product Details