Assay Design
Gene Transcripts
- Gene Symbol
- twk-42
- Entrez Gene ID
-
180163
- Gene Name
- TWiK family of potassium channels
- Gene Aliases
-
CELE_W06D12.2
- Chromosome Location
-
Chr.V: 17726791 - 17732616 on Build WBcel235
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_075082.7 |
NP_507483.1 |
6 - 7 |
1095 |
158 |
View Full Product Details