Assay Design
Gene Transcripts
- Gene Symbol
- cutl-4
- Entrez Gene ID
-
189246
- Gene Name
- ZP domain-containing protein
- Gene Aliases
-
CELE_W06D12.1
- Chromosome Location
-
Chr.V: 17733546 - 17739901 on Build WBcel235
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_075083.6 |
NP_507484.3 |
7 - 8 |
1191 |
90 |
View Full Product Details