Assay Design



Gene Transcripts

Gene Symbol
dpy-21
Entrez Gene ID
180176
Gene Name
Lysine-specific demethylase 9
Gene Aliases
CELE_Y59A8B.1
Chromosome Location
Chr.V: 17936282 - 17949193 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001029095.4 NP_001024266.1 8 - 9 4510 69
NR_002407.1 7 - 8 933 69

View Full Product Details