Assay Design



Gene Transcripts

Gene Symbol
nhr-79
Entrez Gene ID
180265
Gene Name
protein-coding
Gene Aliases
CELE_T26H2.9
Chromosome Location
Chr.V: 19207676 - 19217180 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001373193.1 NP_001360177.1 2 - 3 169 125
NM_075333.8 NP_507734.4 6 - 7 963 125

View Full Product Details