Assay Design
Gene Transcripts
- Gene Symbol
- srr-2
- Entrez Gene ID
-
188953
- Gene Name
- Serpentine Receptor, class R
- Gene Aliases
-
CELE_T26H2.8
- Chromosome Location
-
Chr.V: 19213667 - 19217176 on Build WBcel235
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_075334.3 |
NP_507735.1 |
2 - 3 |
476 |
90 |
- Gene Symbol
- nhr-79
- Entrez Gene ID
-
180265
- Gene Name
- protein-coding
- Gene Aliases
-
CELE_T26H2.9
- Chromosome Location
-
Chr.V: 19207676 - 19217180 on Build WBcel235
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_075333.8 |
NP_507734.4 |
2 - 3 |
480 |
90 |
View Full Product Details