Assay Design



Gene Transcripts

Gene Symbol
str-81
Entrez Gene ID
188951
Gene Name
Seven TM Receptor
Gene Aliases
CELE_T26H2.6
Chromosome Location
Chr.V: 19225521 - 19251083 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_075336.2 NP_507737.1 4 - 5 777 134
NR_102121.1 6 - 7 991 134

View Full Product Details