Assay Design
Gene Transcripts
- Gene Symbol
- fbxb-2
- Entrez Gene ID
-
188948
- Gene Name
- F-box domain-containing protein
- Gene Aliases
-
CELE_T26H2.3
- Chromosome Location
-
Chr.V: 19235995 - 19237304 on Build WBcel235
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_075339.2 |
NP_507740.1 |
1 - 2 |
91 |
77 |
View Full Product Details