Assay Design
Gene Transcripts
- Gene Symbol
- fbxb-57
- Entrez Gene ID
-
186292
- Gene Name
- F-box domain-containing protein
- Gene Aliases
-
CELE_F55C9.4
- Chromosome Location
-
Chr.V: 19288755 - 19289938 on Build WBcel235
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_075352.3 |
NP_507753.1 |
1 - 2 |
179 |
76 |
View Full Product Details