Assay Design



Gene Transcripts

Gene Symbol
gly-4
Entrez Gene ID
180302
Gene Name
protein-coding
Gene Aliases
CELE_Y116F11B.12
Chromosome Location
Chr.V: 19881372 - 19902647 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001029045.5 NP_001024216.1 9 - 10 1864 70
NM_001269959.3 NP_001256888.1 6 - 7 1401 70
NM_075449.9 NP_507850.2 6 - 7 1403 70
GenBank mRNA AF031834.1 - 6 - 7 1416 70

View Full Product Details