Assay Design
Gene Transcripts
- Gene Symbol
- sec-23
- Entrez Gene ID
-
180317
- Gene Name
- Protein transport protein SEC23
- Gene Aliases
-
CELE_Y113G7A.3
- Chromosome Location
-
Chr.V: 20091263 - 20105693 on Build WBcel235
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_075476.7 |
NP_507877.1 |
8 - 9 |
1602 |
65 |
View Full Product Details