Assay Design
Gene Transcripts
- Gene Symbol
- mlt-4
- Entrez Gene ID
-
180329
- Gene Name
- ANK_REP_REGION domain-containing protein
- Gene Aliases
-
CELE_ZC15.7
- Chromosome Location
-
Chr.V: 20281387 - 20289417 on Build WBcel235
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_075511.6 |
NP_507912.2 |
8 - 9 |
1574 |
68 |
GenBank mRNA |
DQ645956.1 |
- |
8 - 9 |
1567 |
68 |
View Full Product Details