Assay Design
Gene Transcripts
- Gene Symbol
- grd-11
- Entrez Gene ID
-
186880
- Gene Name
- GRounDhog (hedgehog-like family)
- Gene Aliases
-
CELE_K02E2.2
- Chromosome Location
-
Chr.V: 20356772 - 20365773 on Build WBcel235
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_075522.4 |
NP_507923.2 |
6 - 7 |
1042 |
90 |
View Full Product Details