Assay Design
Gene Transcripts
- Gene Symbol
- srxa-15
- Entrez Gene ID
-
189895
- Gene Name
- protein-coding
- Gene Aliases
-
CELE_Y44A6B.2
- Chromosome Location
-
Chr.V: 20642196 - 20644380 on Build WBcel235
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_075588.3 |
NP_507989.2 |
4 - 5 |
582 |
135 |
View Full Product Details