Assay Design
Gene Transcripts
- Gene Symbol
- ife-2
- Entrez Gene ID
-
180393
- Gene Name
- Eukaryotic translation initiation factor 4E-2
- Gene Aliases
-
CELE_R04A9.4
- Chromosome Location
-
Chr.X: 381338 - 382709 on Build WBcel235
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_075693.5 |
NP_508094.1 |
2 - 3 |
258 |
97 |
View Full Product Details