Assay Design
Gene Transcripts
- Gene Symbol
- che-2
- Entrez Gene ID
-
180401
- Gene Name
- Uncharacterized protein
- Gene Aliases
-
CELE_F38G1.1
- Chromosome Location
-
Chr.X: 481450 - 486504 on Build WBcel235
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_075705.6 |
NP_508106.1 |
12 - 13 |
2095 |
96 |
GenBank mRNA |
AJ011523.1 |
- |
12 - 13 |
2117 |
96 |
View Full Product Details