Assay Design
Gene Transcripts
- Gene Symbol
- mrp-2
- Entrez Gene ID
-
180408
- Gene Name
- Uncharacterized protein
- Gene Aliases
-
CELE_F57C12.4
- Chromosome Location
-
Chr.X: 563711 - 572701 on Build WBcel235
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_075720.7 |
NP_508121.1 |
10 - 11 |
3115 |
141 |
View Full Product Details