Assay Design
Gene Transcripts
- Gene Symbol
- arr-1
- Entrez Gene ID
-
180446
- Gene Name
- Beta-arrestin arr-1
- Gene Aliases
-
CELE_F53H8.2
- Chromosome Location
-
Chr.X: 950773 - 954233 on Build WBcel235
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_075782.7 |
NP_508183.1 |
8 - 9 |
1147 |
71 |
View Full Product Details