Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001380900.1 | NP_001367482.1 | 13 - 14 | 2181 | 77 |
NM_001380901.2 | NP_001367480.1 | 16 - 17 | 2792 | 77 | |
NM_001380902.1 | NP_001367483.1 | 13 - 14 | 2187 | 77 | |
NM_001380903.1 | NP_001367481.1 | 16 - 17 | 2804 | 77 | |
NM_001380904.1 | NP_001367484.1 | 14 - 15 | 2556 | 77 |