Assay Design



Gene Transcripts

Gene Symbol
dgk-1
Entrez Gene ID
180452
Gene Name
protein-coding
Gene Aliases
CELE_C09E10.2
Chromosome Location
Chr.X: 983905 - 991462 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001380901.2 NP_001367480.1 8 - 9 1107 108
NM_001380903.1 NP_001367481.1 8 - 9 1119 108
NM_001380904.1 NP_001367484.1 6 - 7 871 108

View Full Product Details