Assay Design
Gene Transcripts
- Gene Symbol
- cutl-21
- Entrez Gene ID
-
180453
- Gene Name
- CUTiclin-Like
- Gene Aliases
-
CELE_F55A4.10
- Chromosome Location
-
Chr.X: 1004337 - 1009422 on Build WBcel235
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_075791.7 |
NP_508192.2 |
2 - 3 |
193 |
79 |
View Full Product Details