Assay Design
Gene Transcripts
- Gene Symbol
- snx-1
- Entrez Gene ID
-
180468
- Gene Name
- PX domain-containing protein
- Gene Aliases
-
CELE_C05D9.1
- Chromosome Location
-
Chr.X: 1128984 - 1133431 on Build WBcel235
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_075815.7 |
NP_508216.1 |
4 - 5 |
919 |
56 |
View Full Product Details