Assay Design



Gene Transcripts

Gene Symbol
pdk-1
Entrez Gene ID
180475
Gene Name
protein-coding
Gene Aliases
CELE_H42K12.1
Chromosome Location
Chr.X: 1321344 - 1327986 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001029571.4 NP_001024742.1 4 - 5 783 71
GenBank mRNA AF130407.1 - 4 - 5 783 71

View Full Product Details