Assay Design
Gene Transcripts
- Gene Symbol
- dpy-3
- Entrez Gene ID
-
180529
- Gene Name
- Col_cuticle_N domain-containing protein
- Gene Aliases
-
CELE_EGAP7.1
- Chromosome Location
-
Chr.X: 2144599 - 2145647 on Build WBcel235
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_075994.6 |
NP_508395.1 |
1 - 2 |
167 |
95 |
View Full Product Details