Assay Design
Gene Transcripts
- Gene Symbol
- hex-1
- Entrez Gene ID
-
180533
- Gene Name
- Beta-hexosaminidase A
- Gene Aliases
-
CELE_T14F9.3
- Chromosome Location
-
Chr.X: 2221150 - 2226620 on Build WBcel235
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_076008.7 |
NP_508409.1 |
5 - 6 |
583 |
158 |
GenBank mRNA |
AM748820.1 |
- |
4 - 5 |
468 |
158 |
View Full Product Details