Assay Design
Gene Transcripts
- Gene Symbol
- hex-1
- Entrez Gene ID
-
180533
- Gene Name
- Beta-hexosaminidase A
- Gene Aliases
-
CELE_T14F9.3
- Chromosome Location
-
Chr.X: 2221150 - 2226620 on Build WBcel235
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_076008.7 |
NP_508409.1 |
8 - 9 |
1205 |
126 |
GenBank mRNA |
AM748820.1 |
- |
7 - 8 |
1090 |
126 |
View Full Product Details