Assay Design
Gene Transcripts
- Gene Symbol
- hum-6
- Entrez Gene ID
-
180539
- Gene Name
- Unconventional myosin heavy chain 6
- Gene Aliases
-
CELE_T10H10.1
- Chromosome Location
-
Chr.X: 2303078 - 2314914 on Build WBcel235
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_076019.5 |
NP_508420.1 |
4 - 5 |
507 |
131 |
View Full Product Details