Assay Design



Gene Transcripts

Gene Symbol
syg-1
Entrez Gene ID
180555
Gene Name
protein-coding
Gene Aliases
CELE_K02E10.8
Chromosome Location
Chr.X: 2509313 - 2515324 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001129687.6 NP_001123159.1 3 - 4 392 85
NM_001373246.4 NP_001360786.1 3 - 4 393 85
NM_076056.8 NP_508457.3 3 - 4 386 85

View Full Product Details