Assay Design
Gene Transcripts
- Gene Symbol
- vang-1
- Entrez Gene ID
-
180579
- Gene Name
- protein-coding
- Gene Aliases
-
CELE_B0410.2
- Chromosome Location
-
Chr.X: 2885858 - 2890681 on Build WBcel235
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_076099.5 |
NP_508500.1 |
2 - 3 |
302 |
58 |
View Full Product Details