Assay Design



Gene Transcripts

Gene Symbol
vang-1
Entrez Gene ID
180579
Gene Name
protein-coding
Gene Aliases
CELE_B0410.2
Chromosome Location
Chr.X: 2885858 - 2890681 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_076099.5 NP_508500.1 3 - 4 432 131
NM_076100.7 NP_508501.1 3 - 4 409 131

View Full Product Details