Assay Design
Gene Transcripts
- Gene Symbol
- twk-16
- Entrez Gene ID
-
180594
- Gene Name
- TWiK family of potassium channels
- Gene Aliases
-
CELE_F52E4.4
- Chromosome Location
-
Chr.X: 3081125 - 3085401 on Build WBcel235
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_076125.4 |
NP_508526.2 |
6 - 7 |
814 |
83 |
GenBank mRNA |
AF083645.1 |
- |
6 - 7 |
806 |
83 |
View Full Product Details