Assay Design



Gene Transcripts

Gene Symbol
spc-1
Entrez Gene ID
180603
Gene Name
protein-coding
Gene Aliases
CELE_K10B3.10
Chromosome Location
Chr.X: 3117439 - 3127079 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001270070.3 NP_001256999.1 9 - 10 6581 77
NM_001270071.5 NP_001257000.1 10 - 11 6578 77

View Full Product Details