Assay Design



Gene Transcripts

Gene Symbol
rgl-1
Entrez Gene ID
180611
Gene Name
protein-coding
Gene Aliases
CELE_F28B4.2
Chromosome Location
Chr.X: 3210468 - 3220367 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001129668.5 NP_001123140.1 4 - 5 397 73
NM_076149.7 NP_508550.1 3 - 4 411 73

View Full Product Details