Assay Design
Gene Transcripts
- Gene Symbol
- pqn-37
- Entrez Gene ID
-
185539
- Gene Name
- Prion-like-(Q/N-rich)-domain-bearing protein
- Gene Aliases
-
CELE_F40F4.8
- Chromosome Location
-
Chr.X: 3247042 - 3250531 on Build WBcel235
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_076153.5 |
NP_508554.3 |
7 - 8 |
1098 |
81 |
View Full Product Details