Assay Design
Gene Transcripts
- Gene Symbol
- sul-1
- Entrez Gene ID
-
180619
- Gene Name
- Putative extracellular sulfatase Sulf-1 homolog
- Gene Aliases
-
CELE_K09C4.8
- Chromosome Location
-
Chr.X: 3267294 - 3270906 on Build WBcel235
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_076159.4 |
NP_508560.1 |
8 - 9 |
1358 |
159 |
View Full Product Details