Assay Design
Gene Transcripts
- Gene Symbol
- dhs-27
- Entrez Gene ID
-
180632
- Gene Name
- Uncharacterized oxidoreductase dhs-27
- Gene Aliases
-
CELE_C04F6.5
- Chromosome Location
-
Chr.X: 3412876 - 3414712 on Build WBcel235
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_076190.5 |
NP_508591.3 |
6 - 7 |
846 |
85 |
View Full Product Details