Assay Design
Gene Transcripts
- Gene Symbol
- tag-52
- Entrez Gene ID
-
180657
- Gene Name
- Putative protein tag-52
- Gene Aliases
-
CELE_C02F12.4
- Chromosome Location
-
Chr.X: 3674041 - 3678557 on Build WBcel235
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_076232.4 |
NP_508633.1 |
1 - 2 |
232 |
148 |
View Full Product Details