Assay Design
Gene Transcripts
- Gene Symbol
- acr-10
- Entrez Gene ID
-
191598
- Gene Name
- AcetylCholine Receptor
- Gene Aliases
-
CELE_R02E12.8
- Chromosome Location
-
Chr.X: 4022267 - 4025641 on Build WBcel235
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_076291.5 |
NP_508692.3 |
5 - 6 |
729 |
134 |
View Full Product Details