Assay Design



Gene Transcripts

Gene Symbol
srv-6
Entrez Gene ID
183713
Gene Name
Serpentine Receptor, class V
Gene Aliases
CELE_C52B9.5
Chromosome Location
Chr.X: 4257061 - 4259321 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_076327.4 NP_508728.3 6 - 7 798 101

View Full Product Details