Assay Design



Gene Transcripts

Gene Symbol
cka-2
Entrez Gene ID
180703
Gene Name
protein-coding
Gene Aliases
CELE_C52B9.1
Chromosome Location
Chr.X: 4286085 - 4291496 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001029309.5 NP_001024480.1 4 - 5 713 103
NM_001029310.6 NP_001024481.1 4 - 5 1828 103

View Full Product Details