Assay Design
Gene Transcripts
- Gene Symbol
- aman-1
- Entrez Gene ID
-
180749
- Gene Name
- Alpha-mannosidase
- Gene Aliases
-
CELE_F55D10.1
- Chromosome Location
-
Chr.X: 4733095 - 4738592 on Build WBcel235
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_076410.4 |
NP_508811.1 |
9 - 10 |
1245 |
93 |
View Full Product Details