Assay Design
Gene Transcripts
- Gene Symbol
- ifa-4
- Entrez Gene ID
-
180765
- Gene Name
- Intermediate filament protein ifa-4
- Gene Aliases
-
CELE_K05B2.3
- Chromosome Location
-
Chr.X: 4912308 - 4915991 on Build WBcel235
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_076435.9 |
NP_508836.5 |
8 - 9 |
1431 |
116 |
View Full Product Details