Assay Design
Gene Transcripts
- Gene Symbol
- hsd-2
- Entrez Gene ID
-
191049
- Gene Name
- 3Beta_HSD domain-containing protein
- Gene Aliases
-
CELE_ZC8.1
- Chromosome Location
-
Chr.X: 5005240 - 5007037 on Build WBcel235
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_076450.7 |
NP_508851.1 |
6 - 7 |
1046 |
71 |
View Full Product Details