Assay Design



Gene Transcripts

Gene Symbol
ets-5
Entrez Gene ID
180778
Gene Name
protein-coding
Gene Aliases
CELE_C42D8.4
Chromosome Location
Chr.X: 5079657 - 5084526 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001277027.4 NP_001263956.1 3 - 4 354 72
NM_001383573.1 3 - 4 391 72

View Full Product Details